missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Tetradecylthiophene 98.0+%, TCI America™
$108.11 - $536.84
Chemical Identifiers
| CAS | 110851-66-6 |
|---|---|
| Molecular Formula | C18H32S |
| Molecular Weight (g/mol) | 280.51 |
| MDL Number | MFCD09750675 |
| InChI Key | CAEIOINMYGTXNS-UHFFFAOYSA-N |
| PubChem CID | 14900816 |
| IUPAC Name | 3-tetradecylthiophene |
| SMILES | CCCCCCCCCCCCCCC1=CSC=C1 |
Chemical Identifiers
| 110851-66-6 | |
| 280.51 | |
| CAEIOINMYGTXNS-UHFFFAOYSA-N | |
| 3-tetradecylthiophene |
| C18H32S | |
| MFCD09750675 | |
| 14900816 | |
| CCCCCCCCCCCCCCC1=CSC=C1 |
Specifications
| 110851-66-6 | |
| C18H32S | |
| 5 g | |
| CCCCCCCCCCCCCCC1=CSC=C1 | |
| 280.51 | |
| 280.51 | |
| Liquid |
| Yellow | |
| MFCD09750675 | |
| CAEIOINMYGTXNS-UHFFFAOYSA-N | |
| 3-tetradecylthiophene | |
| 14900816 | |
| ≥98.0% (GC) | |
| 3-Tetradecylthiophene |
Safety and Handling
TSCA : No