missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 110°C |
| Quantity | 5 mL |
| Formula Weight | 190.36 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol |
Safety and Handling
| TSCA | No |