missing translation for 'onlineSavingsMsg'
Learn More

3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 96.0+%, TCI America™

Catalog No. B55551ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55551ML 1 mL
B55555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55551ML Supplier TCI America Supplier No. B55551ML
Only null left

Chemical Identifiers

CAS 73842-99-6
Molecular Formula C9H22O2Si
Molecular Weight (g/mol) 190.36
MDL Number MFCD00187968
InChI Key NETUFVYVNJNFMU-UHFFFAOYSA-N
PubChem CID 4077791
IUPAC Name 3-[(tert-butyldimethylsilyl)oxy]propan-1-ol
SMILES CC(C)(C)[Si](C)(C)OCCCO

Specifications

Color Colorless
Boiling Point 110°C
Quantity 1 mL
Formula Weight 190.36
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol