missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyl)-9H-carbazole 95.0+%, TCI America™

Catalog No. b5053200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5053200MG 200 mg
B50531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5053200MG Supplier TCI America Supplier No. B5053200MG
Only null left

Chemical Identifiers

CAS 22401-74-7
Molecular Formula C16H17N
Molecular Weight (g/mol) 223.319
MDL Number MFCD12024281
InChI Key TYXSZNGDCCGIBO-UHFFFAOYSA-N
PubChem CID 12117342
IUPAC Name 3-tert-butyl-9H-carbazole
SMILES CC(C)(C)C1=CC2=C(C=C1)NC3=CC=CC=C32

Specifications

Melting Point 153°C
Color White
Quantity 200 mg
Formula Weight 223.32
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butyl)-9H-carbazole