missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7119012
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13585G 5 g
B135825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13585G Supplier TCI America Supplier No. B13585G
Only null left

Chemical Identifiers

CAS 99724-19-3
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00059040
InChI Key DQQJBEAXSOOCPG-UHFFFAOYSA-N
Synonym 3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine
PubChem CID 2757234
IUPAC Name tert-butyl N-pyrrolidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butoxycarbonylamino)pyrrolidine