missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butoxycarbonylamino)-1-propanol 96.0+%, TCI America™

Catalog No. H09005ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09005ML 5 mL
H090025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09005ML Supplier TCI America Supplier No. H09005ML
Only null left

Chemical Identifiers

CAS 58885-58-8
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD00191883
InChI Key XDJCYKMWJCYQJM-UHFFFAOYSA-N
Synonym 3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, 3-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 3838134
IUPAC Name tert-butyl N-(3-hydroxypropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCO

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 175.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(tert-Butoxycarbonylamino)-1-propanol