missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Sulfopropyl Methacrylate Potassium Salt 97.0+%, TCI America™
$221.83 - $1163.17
Chemical Identifiers
| CAS | 31098-21-2 |
|---|---|
| Molecular Formula | C7H11KO5S |
| Molecular Weight (g/mol) | 246.318 |
| MDL Number | MFCD00010139 |
| InChI Key | PNOXUQIZPBURMT-UHFFFAOYSA-M |
| Synonym | Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt |
| PubChem CID | 23682747 |
| IUPAC Name | potassium;3-(2-methylprop-2-enoyloxy)propane-1-sulfonate |
| SMILES | CC(=C)C(=O)OCCCS(=O)(=O)[O-].[K+] |
Chemical Identifiers
| 31098-21-2 | |
| 246.318 | |
| PNOXUQIZPBURMT-UHFFFAOYSA-M | |
| 23682747 | |
| CC(=C)C(=O)OCCCS(=O)(=O)[O-].[K+] |
| C7H11KO5S | |
| MFCD00010139 | |
| Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt | |
| potassium;3-(2-methylprop-2-enoyloxy)propane-1-sulfonate |
Specifications
| 31098-21-2 | |
| White | |
| MFCD00010139 | |
| Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt | |
| CC(=C)C(=O)OCCCS(=O)(=O)[O-].[K+] | |
| 246.318 | |
| 246.32 | |
| Crystalline Powder |
| 295°C | |
| C7H11KO5S | |
| 25 g | |
| PNOXUQIZPBURMT-UHFFFAOYSA-M | |
| potassium;3-(2-methylprop-2-enoyloxy)propane-1-sulfonate | |
| 23682747 | |
| ≥97.0% (HPLC,T) | |
| 3-Sulfopropyl Methacrylate Potassium Salt |
Safety and Handling
EINECSNumber : (2)-1613&(2)-2829
TSCA : Yes