Learn More
3-Sulfolene, 98%
CAS: 77-79-2 | C4H6O2S | 118.15 g/mol
Supplier: Thermo Scientific Chemicals A1388736
Description
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.
Solubility
Soluble in water, ethanol, benzene, ether.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
| 3-Sulfolene | |
| 63°C to 67°C | |
| 112°C (233°F) | |
| MFCD00005481 | |
| 107004 | |
| 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569 | |
| MBDNRNMVTZADMQ-UHFFFAOYSA-N | |
| 2,5-dihydrothiophene 1,1-dioxide | |
| 6498 | |
| 98% |
| 77-79-2 | |
| 1.314 | |
| C4H6O2S | |
| 500 g | |
| 14,8956 | |
| Soluble in water,ethanol,benzene,ether. | |
| C1C=CCS1(=O)=O | |
| 118.15 | |
| 118.15 |
Chemical Identifiers
| 77-79-2 | |
| 118.15 | |
| MBDNRNMVTZADMQ-UHFFFAOYSA-N | |
| 6498 | |
| C1C=CCS1(=O)=O |
| C4H6O2S | |
| MFCD00005481 | |
| 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569 | |
| 2,5-dihydrothiophene 1,1-dioxide |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 201-059-7
RTECSNumber : XM9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only