missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfolene, 98%

Catalog No. AAA1388722
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1388722 100 g
AAA1388736 500 g
AAA138870B 1000 g
AAA138870I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1388722 Supplier Thermo Scientific Chemicals Supplier No. A1388722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77-79-2 | C4H6O2S | 118.15 g/mol

It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

Solubility
Soluble in water, ethanol, benzene, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.

Chemical Identifiers

CAS 77-79-2
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00005481
InChI Key MBDNRNMVTZADMQ-UHFFFAOYSA-N
Synonym 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
PubChem CID 6498
IUPAC Name 2,5-dihydrothiophene 1,1-dioxide
SMILES C1C=CCS1(=O)=O

Specifications

Melting Point 63°C to 67°C
Density 1.314
Flash Point 112°C (233°F)
Quantity 100 g
Beilstein 107004
Merck Index 14,8956
Solubility Information Soluble in water,ethanol,benzene,ether.
Formula Weight 118.15
Percent Purity 98%
Chemical Name or Material 3-Sulfolene
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 201-059-7
RTECSNumber XM9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only