Learn More
Poltex 3 Ring Binder Holder with Sign Holder
Catalog No. 22000736
Encompass
Change view
Click to view available options
Mounting Type:
Magnet
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22000736 Wall Mount
22000734 Magnet
22000735 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22000736 Supplier Poltex Supplier No. BINDRSIGW
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides convenient storage for 3-ring binder, logbook, or other documents where they are most needed.
  • Front sign sleeve holds an 8.5 x 11 in. sheet of paper.
  • Holder can be mounted opening left, right, or up.
  • Choose from wall mount (screws/anchors included), magnet mount (powerful magnets included), or VHB Tape (very high bond tape included).
  • Durable, cleanable PETG construction.
Specifications
Color Clear
Material PETG
Mounting Type Wall Mount