missing translation for 'onlineSavingsMsg'
Learn More

3-Quinuclidinone hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. p-7021118
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1332014 25 g
AAA1332022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1332014 Supplier Thermo Scientific Chemicals Supplier No. A1332014
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1193-65-3
Molecular Formula C7H12ClNO
Molecular Weight (g/mol) 161.629
MDL Number MFCD00137391
InChI Key RFDPHKHXPMDJJD-UHFFFAOYSA-N
Synonym 3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
PubChem CID 102019
IUPAC Name 1-azabicyclo[2.2.2]octan-3-one;hydrochloride
SMILES C1CN2CCC1C(=O)C2.Cl

Specifications

Melting Point ∼300°C (decomposition)
Quantity 25 g
Beilstein 3695039
Sensitivity Hygroscopic
Formula Weight 161.63
Percent Purity ≥98%
Chemical Name or Material 3-Quinuclidinone hydrochloride
EINECSNumber 214-776-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only