missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Quinolinecarboxylic Acid 98.0+%, TCI America™
$70.84 - $238.60
Chemical Identifiers
| CAS | 6480-68-8 |
|---|---|
| Molecular Formula | C10H7NO2 |
| Molecular Weight (g/mol) | 173.171 |
| MDL Number | MFCD00006770 |
| InChI Key | DJXNJVFEFSWHLY-UHFFFAOYSA-N |
| Synonym | 3-quinolinecarboxylic acid, 3-quinoline carboxylic acid, 3-quinolinecarboxylicacid, pubchem7517, 3-quinolylcarboxylic acid, acmc-1bfc6, 3-quinolinecarboxylic acid 1g, 3-quinolinecarboxylic acid, purum hplc, q3c |
| PubChem CID | 80971 |
| IUPAC Name | quinoline-3-carboxylic acid |
| SMILES | C1=CC=C2C(=C1)C=C(C=N2)C(=O)O |
Chemical Identifiers
| 6480-68-8 | |
| 173.171 | |
| DJXNJVFEFSWHLY-UHFFFAOYSA-N | |
| 80971 | |
| C1=CC=C2C(=C1)C=C(C=N2)C(=O)O |
| C10H7NO2 | |
| MFCD00006770 | |
| 3-quinolinecarboxylic acid, 3-quinoline carboxylic acid, 3-quinolinecarboxylicacid, pubchem7517, 3-quinolylcarboxylic acid, acmc-1bfc6, 3-quinolinecarboxylic acid 1g, 3-quinolinecarboxylic acid, purum hplc, q3c | |
| quinoline-3-carboxylic acid |
Specifications
| 6480-68-8 | |
| White-Yellow | |
| MFCD00006770 | |
| 3-quinolinecarboxylic acid, 3-quinoline carboxylic acid, 3-quinolinecarboxylicacid, pubchem7517, 3-quinolylcarboxylic acid, acmc-1bfc6, 3-quinolinecarboxylic acid 1g, 3-quinolinecarboxylic acid, purum hplc, q3c | |
| C1=CC=C2C(=C1)C=C(C=N2)C(=O)O | |
| 173.171 | |
| 173.17 | |
| Crystalline Powder |
| 280°C | |
| C10H7NO2 | |
| 1 g | |
| DJXNJVFEFSWHLY-UHFFFAOYSA-N | |
| quinoline-3-carboxylic acid | |
| 80971 | |
| ≥98.0% (HPLC,T) | |
| 3-Quinolinecarboxylic Acid |
Safety and Handling
TSCA : No