Learn More
3-Pyrroline, tech. 85% (remainder pyrrolidine)
CAS: 109-96-6 | C4H7N | 69.107 g/mol
Supplier: Thermo Scientific Chemicals H2607206
Description
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.
Solubility
Soluble in alcohol and ether. Miscible with water.
Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
Specifications
| 3-Pyrroline | |
| 0.902 | |
| −18°C (0°F) | |
| (remainder pyrrolidine) | |
| 1.469 | |
| 5 g | |
| 103173 | |
| 14,8017 | |
| Soluble in alcohol and ether. Miscible with water. | |
| C1C=CCN1 | |
| 69.107 | |
| CHEBI:20198 | |
| 85% |
| 109-96-6 | |
| 90°C to 91°C | |
| Unpleasant | |
| C4H7N | |
| MFCD00005213 | |
| UN2733 | |
| Air sensitive; Hygroscopic | |
| 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole | |
| JVQIKJMSUIMUDI-UHFFFAOYSA-N | |
| 2,5-dihydro-1H-pyrrole | |
| 66059 | |
| 69.11 | |
| Technical |
Chemical Identifiers
| 109-96-6 | |
| 69.107 | |
| JVQIKJMSUIMUDI-UHFFFAOYSA-N | |
| CHEBI:20198 | |
| C1C=CCN1 |
| C4H7N | |
| MFCD00005213 | |
| 66059 | |
| 2,5-dihydro-1H-pyrrole |
Safety and Handling
GHS H Statement
H225-H314-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 203-723-1
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only