Learn More
3-Pyrroline, 96%
CAS: 109-96-6 | C4H7N | 69.107 g/mol
$204.78 - $706.12
Chemical Identifiers
| CAS | 109-96-6 |
|---|---|
| Molecular Formula | C4H7N |
| Molecular Weight (g/mol) | 69.107 |
| MDL Number | MFCD00005213 |
| InChI Key | JVQIKJMSUIMUDI-UHFFFAOYSA-N |
| PubChem CID | 66059 |
| ChEBI | CHEBI:20198 |
| IUPAC Name | 2,5-dihydro-1H-pyrrole |
| SMILES | C1C=CCN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAL1634903
|
Thermo Scientific Chemicals
L1634903 |
1 g |
Each for $204.78
|
|
|||||
|
AAL1634906
|
Thermo Scientific Chemicals
L1634906 |
5 g |
Each for $706.12
|
|
|||||
Description
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.
Solubility
Fully misicble in water.
Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, heat.
Chemical Identifiers
| 109-96-6 | |
| 69.107 | |
| JVQIKJMSUIMUDI-UHFFFAOYSA-N | |
| CHEBI:20198 | |
| C1C=CCN1 |
| C4H7N | |
| MFCD00005213 | |
| 66059 | |
| 2,5-dihydro-1H-pyrrole |
Specifications
| 109-96-6 | |
| 90°C to 91°C | |
| C4H7N | |
| MFCD00005213 | |
| UN2733 | |
| Hygroscopic | |
| 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole | |
| JVQIKJMSUIMUDI-UHFFFAOYSA-N | |
| 2,5-dihydro-1H-pyrrole | |
| 66059 | |
| 69.11 | |
| 3-Pyrroline |
| 0.902 | |
| −18°C (0°F) | |
| 1.469 | |
| 1 g | |
| 103173 | |
| 14,8017 | |
| Fully misicble in water. | |
| C1C=CCN1 | |
| 69.107 | |
| CHEBI:20198 | |
| 96% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 203-723-1
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon; Store under Nitrogen
RUO – Research Use Only