missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrroline, 96%

Catalog No. AAL1634903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1634903 1 g
AAL1634906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1634903 Supplier Thermo Scientific Chemicals Supplier No. L1634903

CAS: 109-96-6 | C4H7N | 69.107 g/mol

3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

Solubility
Fully misicble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, heat.

Chemical Identifiers

CAS 109-96-6
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00005213
InChI Key JVQIKJMSUIMUDI-UHFFFAOYSA-N
PubChem CID 66059
ChEBI CHEBI:20198
IUPAC Name 2,5-dihydro-1H-pyrrole
SMILES C1C=CCN1

Specifications

Density 0.902
Boiling Point 90°C to 91°C
Flash Point −18°C (0°F)
Refractive Index 1.469
Quantity 1 g
UN Number UN2733
Beilstein 103173
Sensitivity Hygroscopic
Merck Index 14,8017
Synonym 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole
Solubility Information Fully misicble in water.
Formula Weight 69.11
Percent Purity 96%
Chemical Name or Material 3-Pyrroline
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 203-723-1
TSCA Yes
Recommended Storage Keep cold; Store under Argon; Store under Nitrogen

RUO – Research Use Only