missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrrolidin-1-ylphenyl)methanol, 97%, Thermo Scientific™

Catalog No. CC38509CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC38509CB 250 mg
CC38509DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC38509CB Supplier Thermo Scientific Chemicals Supplier No. CC38509CB

Chemical Identifiers

CAS 859850-72-9
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD07772826
InChI Key MYIYSGIMXUQECR-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylphenyl methanol, 3-pyrrolidin-1-yl phenyl methanol, 3-1-pyrrolidinyl benzyl alcohol, 3-pyrrolidinylphenyl methan-1-ol, benzenemethanol,3-1-pyrrolidinyl
PubChem CID 7162062
IUPAC Name (3-pyrrolidin-1-ylphenyl)methanol
SMILES C1CCN(C1)C2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material (3-Pyrrolidin-1-ylphenyl)methanol
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.