missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-2-ylbenzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC56301DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC56301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

579476-26-9
200.197
KLTNGIZIUGYITR-UHFFFAOYSA-N
22317373
C1=CC(=CC(=C1)C(=O)O)C2=NC=CC=N2
C11H8N2O2
MFCD05864812
3-pyrimidin-2-yl-benzoic acid, 3-pyrimidin-2-yl benzoic acid, 2-3-carboxyphenyl pyrimidine, 3-2-pyrimidinyl benzoic acid, benzoic acid,3-2-pyrimidinyl, benzoic acid, 3-2-pyrimidinyl, 3-pyrimidin-2-yl benzoicacid
3-pyrimidin-2-ylbenzoic acid

Specifications

3-Pyrimidin-2-ylbenzoic acid
96.5
97%
C11H8N2O2
MFCD05864812
KLTNGIZIUGYITR-UHFFFAOYSA-N
3-pyrimidin-2-ylbenzoic acid
22317373
97%
579476-26-9
100.0
Amber Glass Bottle
1g
3-pyrimidin-2-yl-benzoic acid, 3-pyrimidin-2-yl benzoic acid, 2-3-carboxyphenyl pyrimidine, 3-2-pyrimidinyl benzoic acid, benzoic acid,3-2-pyrimidinyl, benzoic acid, 3-2-pyrimidinyl, 3-pyrimidin-2-yl benzoicacid
C1=CC(=CC(=C1)C(=O)O)C2=NC=CC=N2
200.197
200.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions