missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyloxyacetic acid, 98%

Catalog No. p-7027773
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2478003 1 g
AAB2478006 5 g
AAB2478014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2478003 Supplier Thermo Scientific Chemicals Supplier No. B2478003

CAS: 86649-57-2 | C7H7NO3 | 153.14 g/mol

3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.0 g/L)

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 86649-57-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD04039772
InChI Key MBEPWLCDXKKANL-UHFFFAOYSA-N
Synonym pyridin-3-yloxy-acetic acid, 2-pyridin-3-yloxy acetic acid, 3-pyridyloxyacetic acid, pyridin-3-yloxy acetic acid, 2-3-pyridyloxy acetic acid, pyridin-3-yloxy-aceticacid, 2-3-pyridinyloxy acetic acid, 2-pyridin-3-yloxy aceticacid, acetic acid, 3-pyridinyloxy, 2-pyridine-3-yl oxyacetic acid
PubChem CID 3159630
IUPAC Name 2-pyridin-3-yloxyacetic acid
SMILES OC(=O)COC1=CN=CC=C1

Specifications

Melting Point 210°C to 212°C
Quantity 1 g
Solubility Information Slightly Soluble in water (2.0g/L)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Pyridyloxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only