missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinepropionic acid, 97%

Catalog No. p-3758734
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390980010 1 g Glass bottle
AC390980050 5 g Glass bottle
AC390980100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC390980010 Supplier Thermo Scientific Chemicals Supplier No. 390980010

CAS: 3724-19-4 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3724-19-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00274197
InChI Key WDGXIUUWINKTGP-UHFFFAOYSA-N
Synonym 3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332
PubChem CID 259624
IUPAC Name 3-pyridin-3-ylpropanoic acid
SMILES C1=CC(=CN=C1)CCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 153°C to 156°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridinepropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only