missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyrid-4-ylbenzoic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Quantity | 250mg |
| Formula Weight | 199.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Pyrid-4-ylbenzoic acid |
Safety and Handling
| Hazard Category | Warning |