missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC50204CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC50204CB 250mg Amber Glass Bottle
CC50204DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC50204CB Supplier Thermo Scientific Chemicals Supplier No. CC50204CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137386-78-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03840110
InChI Key YGXYNKLOBXMTGL-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy benzaldehyde, 3-pyrid-2-yloxy benzaldehyde, 3-2-pyridinyloxy-benzaldehyde, 3-2-pyridyloxy benzaldehyde, 3-2-pyridinyloxy benzaldehyde
PubChem CID 2735363
IUPAC Name 3-pyridin-2-yloxybenzaldehyde
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.