missing translation for 'onlineSavingsMsg'
Learn More

3-Propoxypyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7135000
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1195100MG 100 mg
P11951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1195100MG Supplier TCI America Supplier No. P1195100MG
Only null left

Chemical Identifiers

CAS 14440-94-9
Molecular Formula C9H11NO3
MDL Number MFCD00191574
Synonym 3-Propoxypicolinic Acid

Specifications

Melting Point 118°C
Color White
Quantity 100 mg
Formula Weight 181.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Propoxypyridine-2-carboxylic Acid