missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzoic acid, 97%, Thermo Scientific™

$263.42 - $263.42

Chemical Identifiers

CAS 77940-94-4
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD06659081
InChI Key FLQRORYAJSTYLT-UHFFFAOYSA-N
Synonym 3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
PubChem CID 2795552
IUPAC Name 3-piperidin-1-ylbenzoic acid
SMILES C1CCN(CC1)C2=CC=CC(=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC34001DA
SDS View Documents
Thermo Scientific Chemicals
CC34001DA
1 g Amber Glass Bottle
Each for $263.42
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77940-94-4
205.257
FLQRORYAJSTYLT-UHFFFAOYSA-N
2795552
C1CCN(CC1)C2=CC=CC(=C2)C(=O)O
C12H15NO2
MFCD06659081
3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
3-piperidin-1-ylbenzoic acid

Specifications

77940-94-4
100.0
Amber Glass Bottle
MFCD06659081
3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
C1CCN(CC1)C2=CC=CC(=C2)C(=O)O
205.257
205.26
3-Piperidinobenzoic acid
96.5
97%
C12H15NO2
1 g
FLQRORYAJSTYLT-UHFFFAOYSA-N
3-piperidin-1-ylbenzoic acid
2795552
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'