missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzoic acid, 97%, Thermo Scientific™

Catalog No. CC34001DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC34001DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC34001DA Supplier Thermo Scientific Chemicals Supplier No. CC34001DA

Chemical Identifiers

CAS 77940-94-4
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD06659081
InChI Key FLQRORYAJSTYLT-UHFFFAOYSA-N
Synonym 3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
PubChem CID 2795552
IUPAC Name 3-piperidin-1-ylbenzoic acid
SMILES C1CCN(CC1)C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 3-Piperidinobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.