missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinemethanol 98.0+%, TCI America™

Catalog No. p-7130700
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07375G 5 g
H073725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07375G Supplier TCI America Supplier No. H07375G
Only null left

Chemical Identifiers

CAS 4606-65-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005997
InChI Key VUNPWIPIOOMCPT-UHFFFAOYSA-N
Synonym 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol
PubChem CID 107308
IUPAC Name piperidin-3-ylmethanol
SMILES C1CC(CNC1)CO

Specifications

Melting Point 60°C
Color White
Boiling Point 107°C
Quantity 5 g
UN Number 3263
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Piperidinemethanol
EINECSNumber (5)-3729
TSCA No