Learn More
3-Piperidinemethanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 168041000
| Quantity | 100g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4606-65-9 | |
| 115.17 | |
| 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol | |
| piperidin-3-ylmethanol |
| C6H13NO | |
| VUNPWIPIOOMCPT-UHFFFAOYSA-N | |
| 107308 | |
| C1CC(CNC1)CO |
Specifications
| 3-Piperidinemethanol | |
| 97.5 | |
| 1.0200g/mL | |
| >112°C | |
| 97.5% min. (GC) | |
| C6H13NO | |
| 21,II,8 | |
| 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol | |
| VUNPWIPIOOMCPT-UHFFFAOYSA-N | |
| piperidin-3-ylmethanol | |
| 107308 | |
| 98% |
| 4606-65-9 | |
| 100.0 | |
| 236.0°C | |
| Authentic | |
| Glass bottle | |
| 100g | |
| 1.02 | |
| Solubility in water: soluble | |
| C1CC(CNC1)CO | |
| 115.17 | |
| 115.17 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Gently wash with ple
GHS Signal Word: Danger
EINECSNumber : 225-013-
Promotions