missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinemethanol, 96%

Catalog No. AAL0426414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0426414 25 g
AAL0426406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0426414 Supplier Thermo Scientific Chemicals Supplier No. L0426414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4606-65-9 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4606-65-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005997
InChI Key VUNPWIPIOOMCPT-UHFFFAOYSA-N
Synonym 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol
PubChem CID 107308
IUPAC Name piperidin-3-ylmethanol
SMILES C1CC(CNC1)CO

Specifications

Melting Point 58°C to 62°C
Density 1.025
Boiling Point 236°C
Flash Point >110°C (230°F)
Refractive Index 1.494
Quantity 25 g
UN Number UN3259
Beilstein 103058
Sensitivity Hygroscopic
Formula Weight 115.18
Percent Purity 96%
Chemical Name or Material 3-Piperidinemethanol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 225-013-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only