missing translation for 'onlineSavingsMsg'
Learn More

3-Picolylamine 99.0+%, TCI America™

Supplier:  TCI America P0762500ML

 View more versions of this product

Catalog No. P0762500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Picolylamine
-21°C
11 to 12 (10% H2O 20°C)
C6H8N2
500 mL
3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
C1=CC(=CN=C1)CN
108.144
108.14
Liquid
3731-52-0
Yellow
74°C
MFCD00006412
2735
HDOUGSFASVGDCS-UHFFFAOYSA-N
pyridin-3-ylmethanamine
31018
≥99.0% (GC)

Chemical Identifiers

3731-52-0
108.144
HDOUGSFASVGDCS-UHFFFAOYSA-N
31018
C1=CC(=CN=C1)CN
C6H8N2
MFCD00006412
3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
pyridin-3-ylmethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6097

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions