missing translation for 'onlineSavingsMsg'
Learn More

3-Picolylamine 99.0+%, TCI America™

Catalog No. P076225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P076225ML 25 mL
P0762100ML 100 mL
P0762500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P076225ML Supplier TCI America Supplier No. P076225ML

Chemical Identifiers

CAS 3731-52-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006412
InChI Key HDOUGSFASVGDCS-UHFFFAOYSA-N
Synonym 3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
PubChem CID 31018
IUPAC Name pyridin-3-ylmethanamine
SMILES C1=CC(=CN=C1)CN

Specifications

Melting Point -21°C
Color Yellow
pH 11 to 12 (10% H2O 20°C)
Boiling Point 74°C
Quantity 25 mL
UN Number 2735
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Picolylamine
EINECSNumber (5)-6097
TSCA Yes