missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline-N-oxide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1003-73-2
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006203
InChI Key DMGGLIWGZFZLIY-UHFFFAOYSA-N
Synonym 3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
PubChem CID 13858
IUPAC Name 3-methyl-1-oxidopyridin-1-ium
SMILES CC1=C[N+](=CC=C1)[O-]
View More Specs

Products 1
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
AC131151000
View Documents Promotion Details
N/A 100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1003-73-2
109.13
DMGGLIWGZFZLIY-UHFFFAOYSA-N
13858
CC1=C[N+](=CC=C1)[O-]
C6H7NO
MFCD00006203
3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
3-methyl-1-oxidopyridin-1-ium

Specifications

1003-73-2
100.0
143°C
98%
MFCD00006203
Solubility in water: freely soluble in water
CC1=C[N+](=CC=C1)[O-]
109.13
109.13
3-Picoline-N-oxide, 98%
97.5
150°C (15.0mmHg)
Authentic
C6H7NO
3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
DMGGLIWGZFZLIY-UHFFFAOYSA-N
3-methyl-1-oxidopyridin-1-ium
13858
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-714-4

RTECSNumber : UT5799000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only