missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyl Chloride 97.0+%, TCI America™

Catalog No. p-7135038
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P126225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P126225G Supplier TCI America Supplier No. P126225G

Chemical Identifiers

CAS 104-52-9
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00001001
InChI Key XZBXAYCCBFTQHH-UHFFFAOYSA-N
Synonym 3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
PubChem CID 7706
IUPAC Name 3-chloropropylbenzene
SMILES C1=CC=C(C=C1)CCCCl

Specifications

Color Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 154.64
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropyl Chloride