missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropiophenone, 98%

Catalog No. AAL0917309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0917309 10 g
AAL0917318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0917309 Supplier Thermo Scientific Chemicals Supplier No. L0917309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1083-30-3 | C15H14O | 210.276 g/mol

3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1083-30-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00039563
InChI Key QGGZBXOADPVUPN-UHFFFAOYSA-N
Synonym dihydrochalcone, 3-phenylpropiophenone, benzylacetophenone, hydrochalcone, 1,3-diphenyl-1-propanone, beta-phenylpropiophenone, hydrocinnamophenone, 1,3-diphenyl-1-oxopropane, phenethyl phenyl ketone, phenyl phenethyl ketone
PubChem CID 64802
ChEBI CHEBI:71231
IUPAC Name 1,3-diphenylpropan-1-one
SMILES C1=CC=C(C=C1)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 68°C to 73°C
Quantity 10 g
Beilstein 1910661
Solubility Information Insoluble in water.
Formula Weight 210.28
Percent Purity 98%
Chemical Name or Material 3-Phenylpropiophenone
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only