missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionic Acid 98.0+%, TCI America™

Catalog No. p-7130391
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018325G 25 g
H0183100G 100 g
H0183500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H018325G Supplier TCI America Supplier No. H018325G
Only null left

Chemical Identifiers

CAS 501-52-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002771
InChI Key XMIIGOLPHOKFCH-UHFFFAOYSA-N
Synonym hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
PubChem CID 107
ChEBI CHEBI:28631
IUPAC Name 3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 48°C
Color White
Boiling Point 280°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylpropionic Acid
EINECSNumber (3)-1751
RTECSNumber DA8600000
TSCA Yes