missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionamide 98.0+%, TCI America™

Catalog No. P18455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18455G 5 g
P184525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18455G Supplier TCI America Supplier No. P18455G
Only null left

Chemical Identifiers

CAS 102-93-2
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00025535
InChI Key VYIBCOSBNVFEIW-UHFFFAOYSA-N
PubChem CID 7625
IUPAC Name 3-phenylpropanamide
SMILES C1=CC=C(C=C1)CCC(=O)N

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylpropionamide
RTECSNumber MW4905500
TSCA No