missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Phenyllactic acid, 98+%

CAS: 828-01-3 | C9H10O3 | 166.176 g/mol

Supplier:  Thermo Scientific Chemicals L0985403

 View more versions of this product

Catalog No. AAL0985403


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-3-Phenyllactic acid
95°C to 98°C
C9H10O3
1 g
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
C1=CC=C(C=C1)CC(C(=O)O)O
166.176
CHEBI:25998
≥98%
828-01-3
Odorless
MFCD00065928
2209791
VOXXWSYKYCBWHO-UHFFFAOYSA-N
2-hydroxy-3-phenylpropanoic acid
3848
166.18

Chemical Identifiers

828-01-3
166.176
VOXXWSYKYCBWHO-UHFFFAOYSA-N
3848
2-hydroxy-3-phenylpropanoic acid
C9H10O3
MFCD00065928
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
CHEBI:25998
C1=CC=C(C=C1)CC(C(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 212-580-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only