missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Phenyllactic acid, 98+%

Catalog No. AAL0985406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0985406 5 g
AAL0985403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0985406 Supplier Thermo Scientific Chemicals Supplier No. L0985406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 828-01-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 828-01-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065928
InChI Key VOXXWSYKYCBWHO-UHFFFAOYSA-N
Synonym dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
PubChem CID 3848
ChEBI CHEBI:25998
IUPAC Name 2-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)O

Specifications

Melting Point 95°C to 98°C
Odor Odorless
Quantity 5 g
Beilstein 2209791
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (+/-)-3-Phenyllactic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 212-580-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only