missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylisoxazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. P23901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23901G 1 g
P23905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23901G Supplier TCI America Supplier No. P23901G

Chemical Identifiers

CAS 14442-12-7
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01936010
InChI Key YGTFDJRCCBKLDM-UHFFFAOYSA-N
Synonym 3-phenylisoxazole-5-carboxylic acid, 3-phenyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-phenyl, 3-phenyl-isoxazole-5-carboxylic acid, phenylisoxazolecarboxylicacid, 5-carboxy-3-phenylisoxazole, 3-phenylisoxazole-5-carboxylic, 3-phenyl-5-isoxazolecarboxylicacid, 3-phenylisoxazole-5-carboxylicacid, 3-phenyl-5-isoxazole carboxylic acid
PubChem CID 737474
IUPAC Name 3-phenyl-1,2-oxazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NOC(=C2)C(=O)O

Specifications

Melting Point 178°C
Color Yellow
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylisoxazole-5-carboxylic Acid