missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylcinnoline-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. KM09072DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM09072DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM09072DA Supplier Thermo Scientific Chemicals Supplier No. KM09072DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10604-21-4
Molecular Formula C15H10N2O2
Molecular Weight (g/mol) 250.257
MDL Number MFCD00219795
InChI Key UGJHDXUWDLKCDG-UHFFFAOYSA-N
Synonym 4-cinnolinecarboxylicacid, 3-phenyl, maybridge1_004872, 3-phenyl-cinnoline-4-carboxylic acid
PubChem CID 5709072
IUPAC Name 3-phenylcinnoline-4-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N=N2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 250.26
Percent Purity 97%
Chemical Name or Material 3-Phenylcinnoline-4-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.