missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenyl-9H-carbazole 98.0+%, TCI America™
$162.17 - $561.08
Chemical Identifiers
| CAS | 103012-26-6 |
|---|---|
| Molecular Formula | C18H13N |
| Molecular Weight (g/mol) | 243.309 |
| MDL Number | MFCD22052094 |
| InChI Key | IAWRFMPNMXEJCK-UHFFFAOYSA-N |
| PubChem CID | 13497473 |
| IUPAC Name | 3-phenyl-9H-carbazole |
| SMILES | C1=CC=C(C=C1)C2=CC3=C(C=C2)NC4=CC=CC=C43 |
Chemical Identifiers
| 103012-26-6 | |
| 243.309 | |
| IAWRFMPNMXEJCK-UHFFFAOYSA-N | |
| 3-phenyl-9H-carbazole |
| C18H13N | |
| MFCD22052094 | |
| 13497473 | |
| C1=CC=C(C=C1)C2=CC3=C(C=C2)NC4=CC=CC=C43 |
Specifications
| 103012-26-6 | |
| C18H13N | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC3=C(C=C2)NC4=CC=CC=C43 | |
| 243.309 | |
| 243.31 | |
| Crystalline Powder |
| 220°C | |
| MFCD22052094 | |
| IAWRFMPNMXEJCK-UHFFFAOYSA-N | |
| 3-phenyl-9H-carbazole | |
| 13497473 | |
| ≥98.0% (GC) | |
| 3-Phenyl-9H-carbazole |
Safety and Handling
TSCA : No