missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyne (stabilized with BHT) 95.0+%, TCI America™

Catalog No. P19561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19561G 1 g
P19565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19561G Supplier TCI America Supplier No. P19561G

Chemical Identifiers

CAS 10147-11-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00134431
InChI Key NGKSKVYWPINGLI-UHFFFAOYSA-N
Synonym 3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
PubChem CID 575753
IUPAC Name prop-2-ynylbenzene
SMILES C#CCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 75°C
Quantity 1 g
UN Number 3295
Formula Weight 116.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propyne (stabilized with BHT)