missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propylboronic acid pinacol ester, 97%

Catalog No. AAH2898503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2898503 1 g
AAH2898506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2898503 Supplier Thermo Scientific Chemicals Supplier No. H2898503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 329685-40-7 | C15H23BO2 | 246.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329685-40-7
Molecular Formula C15H23BO2
Molecular Weight (g/mol) 246.16
MDL Number MFCD09953505
InChI Key HRZOKAQQKKQUME-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-phenylpropyl-1,3,2-dioxaborolane, 3-phenyl-1-propylboronic acid pinacol ester, 2-3-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 15605871
IUPAC Name 4,4,5,5-tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(CCCC2=CC=CC=C2)OC1(C)C

Specifications

Quantity 1 g
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material 3-Phenyl-1-propylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only