missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol 98.0+%, TCI America™

Catalog No. p-7134492
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021425ML 25 mL
P0214500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P021425ML Supplier TCI America Supplier No. P021425ML
Only null left

Chemical Identifiers

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO

Specifications

Color Yellow
Boiling Point 236°C
Quantity 25 mL
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propanol
EINECSNumber (3)-1034
RTECSNumber UB8970000
TSCA Yes