missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol, 99%

Catalog No. p-7020909
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302230 250 g
AAA130220B 1000 g
AAA130220I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1302230 Supplier Thermo Scientific Chemicals Supplier No. A1302230
Only null left

CAS: 122-97-4 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3-Phenyl-1-propanol has a natural fragrance with antimicrobial properties against bacteria and molds. It is used in combination with Heliotropin or Piperonal as a preservative for cosmetic products. Its ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth and lily of the valley because of the balsamic odor character. It is also a useful biochemical for proteomics research.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO

Specifications

Melting Point -18°C
Density 1.004
Boiling Point 235°C
Flash Point 109°C (228°F)
Refractive Index 1.526
Linear Formula C6H5(CH2)3OH
Quantity 250 g
Beilstein 1857542
Solubility Information Not miscible in water.
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 3-Phenyl-1-propanol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 204-587-6
RTECSNumber UB8970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only