missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propanol, 98%

Catalog No. p-183424
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161312500 250 g Glass bottle
AC161310010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161312500 Supplier Thermo Scientific Chemicals Supplier No. 161312500
Only null left

CAS: 122-97-4 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-97-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002950
InChI Key VAJVDSVGBWFCLW-UHFFFAOYSA-N
Synonym 3-phenyl-1-propanol, benzenepropanol, hydrocinnamyl alcohol, 3-phenylpropyl alcohol, 3-phenylpropanol, 3-benzenepropanol, phenylpropyl alcohol, hydrocinnamic alcohol, 3-phenyl-n-propanol, 3-hydroxypropyl benzene
PubChem CID 31234
IUPAC Name 3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -18°C
Color Colorless
Density 1.0400g/mL
Boiling Point 235°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.527
Linear Formula C6H5(CH2)3OH
Quantity 250 g
Beilstein 06, 503
Specific Gravity 1.04
Solubility Information Solubility in water: 10.3g/L water (20°C). Other solubilities: soluble in methanol,cyclohexane,benzene and,ethylacetate
Viscosity 20 mPa.s (20°C)
Formula Weight 136.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-587-6
RTECSNumber UB8970000
TSCA TSCA

RUO – Research Use Only