missing translation for 'onlineSavingsMsg'
Learn More

3 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet, 30 microns, BASF Catalyst, Spectrum™ Chemical

Catalog No. 18607930
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18607930 5 g
18607931 25 g
18607932 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607930 Supplier Spectrum Chemical Mfg Cor Supplier No. SP1045G
Only null left

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
IUPAC Name palladium
SMILES [Pd]

Specifications

Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.