missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyn-1-ol, 99%

Catalog No. p-179674
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197330050 5 g Glass bottle
AC197330250 25 g Glass bottle
AC197331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC197330050 Supplier Thermo Scientific Chemicals Supplier No. 197330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10229-10-4 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10229-10-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002956
InChI Key IDYNOORNKYEHHO-UHFFFAOYSA-N
Synonym 3-pentyn-1-ol, 3-pentynol, acmc-1bxad
PubChem CID 66295
IUPAC Name pent-3-yn-1-ol
SMILES CC#CCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.9100g/mL
Boiling Point 154.0°C to 157.0°C
Flash Point 54°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.4570
Linear Formula CH3C≡CCH2CH2OH
Quantity 5 g
Beilstein 01,IV,2227
Specific Gravity 0.91
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material 3-Pentyn-1-ol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 233-550-7
TSCA TSCA

RUO – Research Use Only