missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyn-1-ol, 98%

Catalog No. p-7043829
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0295103 1 g
AAL0295106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0295103 Supplier Thermo Scientific Chemicals Supplier No. L0295103
Only null left

CAS: 10229-10-4 | C5H8O | 84.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10229-10-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00002956
InChI Key IDYNOORNKYEHHO-UHFFFAOYSA-N
Synonym 3-pentyn-1-ol, 3-pentynol, acmc-1bxad
PubChem CID 66295
IUPAC Name pent-3-yn-1-ol
SMILES CC#CCCO

Specifications

Density 0.912
Boiling Point 154°C to 157°C
Flash Point 54°C (129°F)
Odor Recognizable
Refractive Index 1.456
Linear Formula CH3C≡CCH2CH2OH
Quantity 1 g
UN Number UN1987
Beilstein 1735967
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 3-Pentyn-1-ol
Show More Show Less
Hazard Category H226-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 233-550-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only