missing translation for 'onlineSavingsMsg'
Learn More

3-Pentylthiophene 97.0+%, TCI America™

Catalog No. p-7134963
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11271G 1 g
P11275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11271G Supplier TCI America Supplier No. P11271G
Only null left

Chemical Identifiers

CAS 102871-31-8
Molecular Formula C9H14S
Molecular Weight (g/mol) 154.271
MDL Number MFCD00143182
InChI Key PIQKSZYJGUXAQF-UHFFFAOYSA-N
PubChem CID 15345429
IUPAC Name 3-pentylthiophene
SMILES CCCCCC1=CSC=C1

Specifications

Color Colorless
Boiling Point 200°C
Quantity 1 g
UN Number 1993
Formula Weight 154.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pentylthiophene