missing translation for 'onlineSavingsMsg'
Learn More

3-Pentyl Acetoacetate 97.0+%, TCI America™

Catalog No. A081325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A081325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A081325ML Supplier TCI America Supplier No. A081325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13562-81-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00059415
InChI Key PFTIWTQFHWICDR-UHFFFAOYSA-N
Synonym Acetoacetic Acid 3-Pentyl Ester, 3-Amyl Acetoacetate, Acetoacetic Acid 3-Amyl Ester
PubChem CID 14677137
IUPAC Name pentan-3-yl 3-oxobutanoate
SMILES CCC(CC)OC(=O)CC(=O)C

Specifications

Color Colorless
Boiling Point 213°C
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pentyl Acetoacetate
EINECSNumber (2)-1505
TSCA No