missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanol, 98+%

Catalog No. AAL0606522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0606522 100 g
AAL0606536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0606522 Supplier Thermo Scientific Chemicals Supplier No. L0606522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 584-02-1 | C5H12O | 88.15 g/mol

3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pentanol is widely used as a flavoring agent. It is biogenic oxygenated volatile organic compound (BOVOC) and is used as a reagent in the synthesis of pure bromopentanes for infrared standards.

Solubility
Soluble in acetone, benzene, ethanol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 584-02-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004574
InChI Key AQIXEPGDORPWBJ-UHFFFAOYSA-N
Synonym 3-pentanol, diethyl carbinol, sec-pentanol, pentanol-3, 3-pentyl alcohol, 3-hydroxypentane, unii-x4elc182i5, 1-ethyl-1-propanol, c2h5 2choh, isoamyl alcohol primary/secondary
PubChem CID 11428
ChEBI CHEBI:77519
IUPAC Name pentan-3-ol
SMILES CCC(CC)O

Specifications

Melting Point -8°C
Density 0.817
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Odor Characteristic
Refractive Index 1.41
Quantity 100 g
UN Number UN1105
Beilstein 1730964
Merck Index 14,7120
Solubility Information Soluble in acetone,benzene,ethanol and diethyl ether.
Formula Weight 88.15
Percent Purity ≥98%
Chemical Name or Material 3-Pentanol
Show More Show Less
Hazard Category H226-H302-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H315-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber 209-526-7
RTECSNumber SA5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only